Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146193.2 | NP_666305.2 | 8 | 2519 | ||
GenBank mRNA | BC031192.1 | - | 7 | 1997 | 5043252 | |
BC072618.1 | - | 8 | 2519 | 6811133 |