Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006205.2 | NP_006196.1 | 2, 3 | 247 | ||
XM_017019431.1 | XP_016874920.1 | 1, 2 | 490 | |||
GenBank mRNA | BC069774.1 | - | 1, 2 | 184 | 7262475 | |
BC069800.1 | - | 1, 2 | 184 | 7262487 | ||
BC093738.1 | - | 1, 2 | 174 | 7939583 | ||
BC093740.1 | - | 1, 2 | 174 | 7939585 | ||
D45399.1 | - | 2, 3 | 225 | |||
EL946662.1 | - | 2, 3 | 251 |