Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153598.2 | NP_705826.1 | 1 | 465 | ||
GenBank mRNA | AK145951.1 | - | 1 | 475 | ||
BC028826.1 | - | 1 | 465 | 5098890 |