Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278698.1 | NP_001265627.1 | 4 | 818 | ||
NM_001278699.1 | NP_001265628.1 | 4 | 827 | |||
NM_001278700.1 | NP_001265629.1 | 4 | 668 | |||
NM_006232.3 | NP_006223.2 | 4 | 818 | |||
XM_006713666.2 | XP_006713729.1 | 5 | 1356 | |||
XM_006713667.2 | XP_006713730.1 | 5 | 1105 | |||
XM_006713668.3 | XP_006713731.1 | 5 | 1365 | |||
XM_006713670.2 | XP_006713733.1 | 5 | 1365 | |||
GenBank mRNA | BC000739.2 | - | 4 | 368 | 3504683 | |
BG529732.1 | - | 4 | 399 | |||
BM541682.1 | - | 4 | 625 | |||
U37689.1 | - | 4 | 473 | |||
Z49199.1 | - | 4 | 386 |