Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176838.2 | NP_789808.1 | 14 | 2252 | ||
GenBank mRNA | AK020578.1 | - | 1 | 80 | ||
AK088755.1 | - | 14 | 2122 | |||
AK168488.1 | - | 13 | 1485 | |||
BC031444.1 | - | 14 | 2252 | 4222237 |