Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254757.1 | NP_001241686.1 | 4 | 428 | ||
NM_001254758.1 | NP_001241687.1 | 4 | 301 | |||
NM_001254759.1 | NP_001241688.1 | 2 | 445 | |||
NM_006278.2 | NP_006269.1 | 4 | 627 | |||
XM_011542959.1 | XP_011541261.1 | 3 | 230 | |||
XM_011542960.2 | XP_011541262.1 | 3 | 230 | |||
XM_017018134.1 | XP_016873623.1 | 4 | 313 | |||
GenBank mRNA | AF516602.1 | - | 2 | 137 | ||
AF516603.1 | - | 2 | 125 | |||
AF516604.1 | - | 2 | 155 | |||
AF525084.1 | - | 2 | 167 | |||
AK021929.1 | - | 2 | 129 | |||
AK291577.1 | - | 3 | 239 | |||
AK314738.1 | - | 3 | 227 | |||
AL520639.3 | - | 4 | 386 | |||
AL529887.3 | - | 4 | 302 | |||
AY040826.1 | - | 3 | 170 | |||
BC010645.1 | - | 4 | 301 | 3449625 | ||
BI259855.1 | - | 4 | 396 | |||
CR456858.1 | - | 3 | 158 | |||
DA210431.1 | - | 4 | 428 | |||
DA841563.1 | - | 2 | 446 | |||
L23767.1 | - | 2 | 167 | |||
X74570.1 | - | 4 | 320 |