Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178936.3 | NP_849267.1 | 8 | 3757 | ||
GenBank mRNA | AK147601.1 | - | 8 | 3804 | ||
BC040365.1 | - | 8 | 3756 | 4223855 |