Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080548.1 | NP_001074017.1 | 3 | 337 | ||
NM_181399.3 | NP_852064.2 | 2 | 176 | |||
GenBank mRNA | AK084045.1 | - | 3 | 160 | ||
AK129037.1 | - | 3 | 189 | |||
AK155073.1 | - | 3 | 338 | |||
BC051184.1 | - | 2 | 179 | |||
BE949333.1 | - | 3 | 426 | |||
BQ938063.1 | - | 2 | 176 |