Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021535.4 | NP_067510.3 | 3 | 448 | ||
GenBank mRNA | AB044414.1 | - | 3 | 377 | ||
AK003493.1 | - | 3 | 435 | |||
AK011140.1 | - | 3 | 447 | |||
AK019174.1 | - | 3 | 407 | |||
AK145982.1 | - | 3 | 447 | |||
AK159635.1 | - | 3 | 435 | |||
AK168813.1 | - | 3 | 442 | |||
BC020034.1 | - | 3 | 390 | |||
BC057446.1 | - | 3 | 409 | 3155760 |