Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023626.4 | NP_076115.3 | 5, 6 | 521 | ||
GenBank mRNA | AK030364.1 | - | 4, 5 | 328 | ||
AK080787.1 | - | 5, 6 | 490 | |||
AK131993.1 | - | 5, 6 | 521 | |||
AK146145.1 | - | 4, 5 | 338 | |||
AY007791.1 | - | 5, 6 | 490 | |||
BC018342.1 | - | 5, 6 | 471 | 4010253 |