Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023799.3 | NP_076288.1 | 16 | 3473 | ||
GenBank mRNA | AK077613.1 | - | 4 | 1995 | ||
AK129188.1 | - | 16 | 3702 | |||
BC054821.1 | - | 16 | 3473 | 6515250 |