Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024266.3 | NP_077228.1 | 3 | 220 | ||
GenBank mRNA | AK012696.1 | - | 3 | 240 | ||
AK077450.1 | - | 3 | 194 | |||
AK169736.1 | - | 3 | 240 | |||
BC002088.1 | - | 3 | 265 | 3487037 | ||
BC025433.1 | - | 798 | ||||
BC027208.1 | - | 3 | 267 | 3982065 | ||
BC079541.1 | - | 3 | 220 | 6824094 | ||
BC092005.1 | - | 3 | 198 | 5683524 | ||
BC094392.1 | - | 3 | 220 | 6407320 |