Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsnaxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024445.4 | NP_077765.1 | 2 | 150 | ||
GenBank mRNA | AF343750.1 | - | 2 | 122 | ||
AK006041.1 | - | 2 | 151 | |||
BC100431.1 | - | 2 | 163 | 30917958 |