Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026998.3 | NP_081274.2 | 5 | 474 | ||
GenBank mRNA | AK011640.1 | - | 6 | 476 | ||
AK013158.1 | - | 5 | 475 | |||
AK182704.1 | - | 4 | 413 | |||
BC025911.1 | - | 5 | 461 | |||
BC061028.1 | - | 6 | 515 | 6594585 |