Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027480.3 | NP_081756.4 | 20 | 3168 | ||
GenBank mRNA | AK017778.1 | - | 4 | 478 | ||
AK173316.1 | - | 18 | 4134 | |||
BC017633.1 | - | 7 | 1839 | |||
BC048236.1 | - | 19 | 3073 | 6313999 |