Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006355.4 | NP_006346.1 | 3, 4 | 999 | ||
XM_005248799.4 | XP_005248856.1 | 3, 4 | 728 | |||
XM_005248800.4 | XP_005248857.1 | 3, 4 | 869 | |||
XM_017010175.1 | XP_016865664.1 | 3, 4 | 888 | |||
GenBank mRNA | AB209906.1 | - | 2, 3 | 841 | ||
AK313248.1 | - | 3, 4 | 886 | |||
BC026930.1 | - | 2, 3 | 845 | 3879491 | ||
BX640949.1 | - | 3, 4 | 916 | |||
CR936707.1 | - | 2, 3 | 710 | |||
U90547.1 | - | 1, 2 | 431 |