Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027669.3 | NP_081945.1 | 8 | 899 | ||
NM_029374.1 | NP_083650.1 | 8 | 899 | |||
GenBank mRNA | AK006458.1 | - | 8 | 837 | ||
AK016798.1 | - | 8 | 827 | |||
AK077091.1 | - | 8 | 900 | |||
BC115618.1 | - | 8 | 810 | 40102763 | ||
BC115619.1 | - | 7 | 945 | 40102765 |