Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253888.1 | NP_001240817.1 | 2 | 454 | ||
NM_001253889.1 | NP_001240818.1 | 2 | 454 | |||
NM_027673.3 | NP_081949.1 | 2 | 454 | |||
GenBank mRNA | AK006144.1 | - | 2 | 429 | ||
AK016890.1 | - | 2 | 456 | |||
AY461664.1 | - | 2 | 346 | |||
DQ397204.1 | - | 2 | 383 | |||
DQ397205.1 | - | 2 | 354 | |||
DQ397206.1 | - | 2 | 354 | |||
EF127819.1 | - | 2 | 456 | |||
EF127820.1 | - | 2 | 456 | |||
EF127821.1 | - | 2 | 455 | |||
EF127822.1 | - | 2 | 456 |