Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253888.1 | NP_001240817.1 | 3 | 978 | ||
NM_001253889.1 | NP_001240818.1 | 3 | 978 | |||
NM_027673.3 | NP_081949.1 | 3 | 948 | |||
GenBank mRNA | AK006144.1 | - | 3 | 953 | ||
AK016890.1 | - | 3 | 950 | |||
AY461664.1 | - | 3 | 840 | |||
DQ397204.1 | - | 3 | 877 | |||
DQ397205.1 | - | 3 | 878 | |||
DQ397206.1 | - | 3 | 878 | |||
EF127819.1 | - | 3 | 950 | |||
EF127820.1 | - | 3 | 980 | |||
EF127821.1 | - | 3 | 979 | |||
EF127822.1 | - | 3 | 980 |