Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111304.1 | NP_001104774.1 | 10 | 2482 | ||
NM_027758.4 | NP_082034.1 | 7 | 1783 | |||
GenBank mRNA | AK049049.1 | - | 7 | 1784 | ||
AK134614.1 | - | 10 | 1759 | |||
AK173050.1 | - | 10 | 1655 | |||
BC057027.1 | - | 6 | 1070 | 6825869 |