Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB093224.1 | - | 6, 7 | 2190 | ||
AF363690.1 | - | 11, 12 | 2920 | |||
AK004685.1 | - | 2, 3 | 319 | |||
AK082152.1 | - | 1, 2 | 82 | |||
AK147276.1 | - | 11, 12 | 3553 | |||
AK147535.1 | - | 12, 13 | 5671 | |||
BC040463.1 | - | 2, 3 | 620 | |||
BC058665.1 | - | 8, 9 | 2438 |