Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025594.1 | NP_001020765.1 | 3 | 198 | ||
NM_027947.2 | NP_082223.2 | 4 | 290 | |||
GenBank mRNA | AK005818.1 | - | 4 | 264 | ||
AK122277.1 | - | 3 | 169 | |||
AK147806.1 | - | 3 | 193 | |||
BC043117.1 | - | 3 | 198 | 6408745 |