Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ist1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028018.2 | NP_082294.1 | 10 | 1600 | ||
GenBank mRNA | AK010262.1 | - | 10 | 1570 | ||
AK050468.1 | - | 9 | 2900 | |||
AK088192.1 | - | 10 | 1555 | |||
AK090233.1 | - | 1 | 608 | |||
AK122215.1 | - | 5 | 6071 | |||
AK152830.1 | - | 10 | 1567 | |||
AK206471.1 | - | 1 | 103 | |||
AK206666.1 | - | 1 | 103 | |||
AK207868.1 | - | 1 | 103 | |||
AK210593.1 | - | 1 | 103 | |||
AK213048.1 | - | 1 | 103 | |||
AK213930.1 | - | 1 | 103 | |||
AK213937.1 | - | 1 | 103 | |||
AK214771.1 | - | 1 | 103 | |||
BC017605.1 | - | 10 | 1566 | 4507347 | ||
BC039052.1 | - | 9 | 2876 | 2631157 |