Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028048.2 | NP_082324.1 | 4 | 1229 | ||
GenBank mRNA | AK007453.1 | - | 4 | 1231 | ||
AK035739.1 | - | 1200 | ||||
AK171131.1 | - | 4 | 1236 | |||
AK181743.1 | - | 2 | 185 | |||
AK186841.1 | - | 2 | 185 | |||
BC004569.1 | - | 4 | 669 | |||
BC019996.1 | - | 4 | 1171 | 4207271 |