Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028094.3 | NP_082370.2 | 1 | 293 | ||
GenBank mRNA | AK008601.1 | - | 1 | 275 | ||
AK050327.1 | - | 1 | 294 | |||
BC025795.1 | - | 1 | 269 | 5133743 |