Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028094.3 | NP_082370.2 | 6 | 1981 | ||
GenBank mRNA | AK008601.1 | - | 6 | 1963 | ||
BC025795.1 | - | 6 | 1957 | 5133743 | ||
BY460703.1 | - | 1 | 198 |