Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028097.3 | NP_082373.1 | 6 | 910 | ||
GenBank mRNA | AK008493.1 | - | 6 | 944 | ||
AK048210.1 | - | 6 | 910 | |||
AK084881.1 | - | 6 | 944 | |||
AK088600.1 | - | 7 | 1080 | |||
AK147765.1 | - | 6 | 943 | |||
AK162563.1 | - | 6 | 943 | |||
AK165916.1 | - | 6 | 972 | |||
AK166237.1 | - | 6 | 996 | |||
BC016240.1 | - | 6 | 928 | 4506688 |