Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252448.1 | NP_001239377.1 | 6 | 464 | ||
NM_001252449.1 | NP_001239378.1 | 4 | 353 | |||
NM_028126.3 | NP_082402.1 | 4 | 353 | |||
GenBank mRNA | AK011451.1 | - | 4 | 299 | ||
AK138358.1 | - | 6 | 465 | |||
AY341872.1 | - | 4 | 312 | |||
AY341878.1 | - | 4 | 312 | |||
BC052063.1 | - | 484 | ||||
BC058517.1 | - | 4 | 254 | 5717255 |