Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028197.2 | NP_082473.2 | 2 | 575 | ||
GenBank mRNA | AK003009.1 | - | 2 | 513 | ||
AK035659.1 | - | 2 | 563 | |||
AK129320.1 | - | 2 | 560 | |||
AK141324.1 | - | 2 | 583 | |||
AK149629.1 | - | 2 | 512 | |||
AK158730.1 | - | 3 | 310 | |||
AK161007.1 | - | 561 | ||||
BC046485.1 | - | 2 | 546 | 5350638 | ||
BC055307.1 | - | 2 | 529 | 6493547 | ||
BC090974.1 | - | 2 | 529 | 6774777 | ||
BY729088.1 | - | 2 | 576 |