Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163608.1 | NP_001157080.1 | 11 | 1219 | ||
NM_028199.3 | NP_082475.3 | 11 | 1198 | |||
GenBank mRNA | AF378760.1 | - | 11 | 1132 | ||
AK010361.1 | - | 11 | 1198 | |||
AK036144.1 | - | 11 | 1173 | |||
AK153684.1 | - | 11 | 1185 | |||
BC114352.2 | - | 11 | 1118 | 40057410 |