Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028300.2 | NP_082576.1 | 5 | 953 | ||
GenBank mRNA | AK012458.1 | - | 5 | 929 | ||
AK080744.1 | - | 3 | 573 | |||
BC019481.1 | - | 5 | 851 | 4190604 | ||
BC039645.1 | - | 5 | 943 | 5373181 |