Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028325.3 | NP_082601.1 | 5 | 2092 | ||
GenBank mRNA | AK012833.1 | - | 5 | 2009 | ||
AV136720.1 | - | 1 | 206 | |||
AY466375.1 | - | 2 | 1858 | |||
BC049071.1 | - | 4 | 1988 | 5701407 | ||
BC055714.1 | - | 4 | 1959 | 6415226 |