Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028360.2 | NP_082636.3 | 10 | 1558 | ||
NM_029704.2 | NP_083980.2 | 9 | 1519 | |||
GenBank mRNA | AK013368.1 | - | 10 | 1512 | ||
AK034075.1 | - | 10 | 1548 | |||
BC026866.1 | - | 8 | 1202 | |||
BC037111.1 | - | 9 | 1282 | 5370464 |