Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127325.1 | NP_001120797.1 | 1149 | |||
NM_006341.3 | NP_006332.3 | 1116 | ||||
GenBank mRNA | AF072933.1 | - | 960 | |||
AF139365.1 | - | 1038 | ||||
AF157482.1 | - | 1116 | ||||
BC015244.1 | - | 930 | 3916742 |