Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028762.1 | NP_083038.1 | 2 | 303 | ||
GenBank mRNA | AK053511.1 | - | 2 | 307 | ||
AK146493.1 | - | 2 | 333 | |||
AK167100.1 | - | 2 | 299 | |||
BC025619.1 | - | 2 | 303 | 5320947 | ||
BC034010.1 | - | 2 | 269 | 4225239 |