Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028803.4 | NP_083079.1 | 4 | 722 | ||
GenBank mRNA | AK009815.1 | - | 4 | 538 | ||
AK050365.1 | - | 4 | 609 | |||
AK050423.1 | - | 4 | 608 | |||
BC017541.1 | - | 4 | 583 | 4526961 |