Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081028.1 | NP_001074497.1 | 16 | 4614 | ||
GenBank mRNA | AK044193.1 | - | 3 | 441 | ||
AK087819.1 | - | 3 | 599 | |||
AK172968.1 | - | 9 | 1357 | |||
BC095969.1 | - | 6 | 1147 | |||
BC116408.1 | - | 1 | 207 | |||
BC128493.1 | - | 1 | 207 | |||
BC144792.1 | - | 3 | 520 | |||
BC145739.1 | - | 3 | 520 | |||
BC152360.1 | - | 1 | 207 | |||
BC152361.1 | - | 1 | 195 |