Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028833.3 | NP_083109.2 | 3, 4 | 296 | ||
GenBank mRNA | AK006472.2 | - | 3, 4 | 172 | ||
AK029333.1 | - | 3, 4 | 195 | |||
BC059223.1 | - | 3, 4 | 296 | 6417270 | ||
BC065128.1 | - | 3, 4 | 183 |