Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028846.5 | NP_083122.1 | 16 | 1925 | ||
GenBank mRNA | AF449715.1 | - | 16 | 1876 | ||
AK029890.1 | - | 1821 | ||||
AK047242.1 | - | 16 | 1844 | |||
AK054279.1 | - | 16 | 1868 | |||
AK087412.1 | - | 16 | 1836 | |||
AK163663.1 | - | 16 | 1835 | |||
AK173083.1 | - | 16 | 1827 | |||
BC019735.1 | - | 2 | 125 | |||
BC037199.1 | - | 10 | 1309 | |||
BC079674.1 | - | 1814 | 30531994 |