Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028887.3 | NP_083163.3 | 32 | 4224 | ||
GenBank mRNA | AK016419.1 | - | 7 | 701 | ||
AK129181.1 | - | 10 | 1221 | |||
BC058618.1 | - | 8 | 876 | |||
BC062946.1 | - | 18 | 2069 | 30357469 | ||
BC076627.1 | - | 25 | 3187 |