Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028946.3 | NP_083222.2 | 5, 6 | 1062 | ||
GenBank mRNA | AK016883.1 | - | 5, 6 | 914 | ||
AK016917.1 | - | 5, 6 | 1042 | |||
AK029525.1 | - | 5, 6 | 1063 | |||
BC025002.1 | - | 4, 5 | 984 | |||
EU846100.1 | - | 4, 5 | 593 |