Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028959.3 | NP_083235.3 | 9 | 1375 | ||
GenBank mRNA | AK011607.1 | - | 4 | 299 | ||
AK017134.1 | - | 9 | 1367 | |||
AK034892.1 | - | 9 | 1368 | |||
AK161033.1 | - | 5 | 700 | |||
AK173189.1 | - | 2 | 3359 | |||
BC060686.1 | - | 8 | 2363 | |||
BC080681.1 | - | 9 | 1091 | |||
BC100549.1 | - | 8 | 1225 |