Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029271.2 | NP_083547.1 | 3 | 581 | ||
GenBank mRNA | AB049650.1 | - | 3 | 612 | ||
AF279255.1 | - | 3 | 558 | |||
AK002753.1 | - | 3 | 582 | |||
AK031791.1 | - | 1 | 2539 | |||
AK078324.1 | - | 3 | 599 | |||
BC037368.1 | - | 2 | 285 | |||
BC052214.1 | - | 3 | 616 | 30018135 |