Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eppin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029325.2 | NP_083601.1 | 1 | 127 | ||
GenBank mRNA | AF346413.2 | - | 1 | 150 | ||
AK006296.1 | - | 1 | 129 | |||
BC048637.1 | - | 1 | 150 | 6771674 |