Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029362.3 | NP_083638.1 | 5 | 1584 | ||
GenBank mRNA | AK156473.1 | - | 5 | 1548 | ||
AK159193.1 | - | 5 | 1547 | |||
AK164550.1 | - | 1474 | ||||
BC059279.1 | - | 5 | 1533 | 4019739 | ||
CF379912.1 | - | 1 | 587 |