Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190989.1 | NP_001177918.1 | 4 | 701 | ||
NM_029561.4 | NP_083837.2 | 4 | 701 | |||
GenBank mRNA | AF411608.1 | - | 4 | 410 | ||
AK002978.1 | - | 2 | 220 | |||
AK031073.1 | - | 4 | 516 | |||
AK049678.1 | - | 4 | 571 | |||
AK078939.1 | - | 4 | 611 | |||
AK084288.1 | - | 3 | 2996 | |||
AK132235.1 | - | 4 | 630 | |||
AK150832.1 | - | 4 | 515 | |||
AK220371.1 | - | 2 | 151 | |||
BC012434.1 | - | 1 | 2194 | |||
BC071237.1 | - | 4 | 620 | 5003738 | ||
BC106125.1 | - | 4 | 628 | |||
BC117075.1 | - | 4 | 476 | 40126394 | ||
BC119092.1 | - | 4 | 476 | 8733841 | ||
CX206555.1 | - | 1 | 65 |