Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139218.1 | NP_631964.1 | 4 | 701 | ||
GenBank mRNA | AF490347.1 | - | 4 | 706 | ||
AK010723.1 | - | 4 | 642 | |||
AK135769.1 | - | 4 | 706 | |||
AK136263.1 | - | 4 | 706 | |||
AY082485.1 | - | 4 | 701 | |||
AY134859.1 | - | 4 | 716 | |||
BC099433.1 | - | 4 | 679 | 30933569 | ||
BC100331.1 | - | 4 | 676 | 30934920 |