Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144535.4 | NP_653118.3 | 6, 7 | 1586 | ||
GenBank mRNA | AK016540.1 | - | 6, 7 | 1535 | ||
AK030928.1 | - | 6, 7 | 1513 | |||
AK041400.1 | - | 6, 7 | 1087 | |||
AK051372.1 | - | 6, 7 | 1533 | |||
AK154166.1 | - | 6, 7 | 1539 | |||
AK164148.1 | - | 6, 7 | 1357 | |||
AK170232.1 | - | 6, 7 | 1363 | |||
AK205351.1 | - | 3, 4 | 247 | |||
AK211028.1 | - | 3, 4 | 239 | |||
BC023094.1 | - | 6, 7 | 1586 | 4017596 |