Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006332.4 | NP_006323.2 | 7 | 804 | ||
GenBank mRNA | AB049659.1 | - | 7 | 792 | ||
AF097362.1 | - | 7 | 775 | |||
AF401212.1 | - | 1 | 49 | |||
AK123477.1 | - | 5 | 1899 | |||
AK291551.1 | - | 7 | 789 | |||
AL558891.3 | - | 7 | 804 | |||
BC021136.1 | - | 7 | 773 | 4863531 | ||
BC031020.1 | - | 7 | 771 | 4690088 | ||
DQ884410.1 | - | 7 | 685 |