Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Purg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098233.1 | NP_001091703.1 | 1 | 592 | ||
NM_152821.2 | NP_690034.1 | 1 | 592 | |||
GenBank mRNA | AF479672.1 | - | 1 | 592 | ||
AF479673.1 | - | 1 | 592 | |||
BC116406.2 | - | 1 | 592 | 40102393 | ||
CD772246.1 | - | 1 | 157 |